Assay Details
Target Gene Details
Entrez Gene ID: | 312 |
Gene Name: | annexin A13 |
Gene Aliases: |
ANX13, ISA |
Location: |
Chr.8:123680794-123737426 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANXA13 | NM_001003954.2 | NP_001003954.1 | ||
| NM_004306.3 | NP_004297.2 | |||
| AJ306450.1 | CAC34622.1 | |||
| AK223374.1 | BAD97094.1 | |||
| AK290281.1 | ||||
| BC125158.1 | ||||
| CR541838.1 | CAG46637.1 | |||
| Z11502.1 | CAA77578.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv612140 | Chr.8:123592519 - 123726378 on Build GRCh38 | Loss |
|
| nsv1032155 | Chr.8:123597462 - 123727319 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map