Assay Details
Target Gene Details
Entrez Gene ID: | 3061 |
Gene Name: | hypocretin receptor 1 |
Gene Aliases: |
OX1R |
Location: |
Chr.1:31617700-31632518 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCRTR1 | NM_001525.2 | 9 | 1626 | NP_001516.2 |
| XM_017001105.1 | 9 | 1894 | XP_016856594.1 | |
| XM_017001106.1 | 9 | 1550 | XP_016856595.1 | |
| XM_017001107.1 | XP_016856596.1 | |||
| AF041243.1 | 7 | 1394 | AAC39601.1 | |
| AK290521.1 | 9 | 1626 | ||
| BC074796.2 | 7 | 1283 | AAH74796.1 | |
| DA803650.1 | 1 | 185 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545951 | Chr.1:31626924 - 31699894 on Build GRCh38 | Gain |
|
| nsv1000976 | Chr.1:31612766 - 31700055 on Build GRCh38 | Loss |
|
| esv2758930 | Chr.1:31209533 - 32000730 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map