Assay Details
Target Gene Details
Entrez Gene ID: | 5524 |
Gene Name: | protein phosphatase 2 phosphatase activator |
Gene Aliases: |
PP2A, PPP2R4, PR53 |
Location: |
Chr.9:129110949-129148946 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPA | NM_001193397.1 | 4 | 341 | NP_001180326.1 |
| NM_001271832.1 | 3 | 468 | NP_001258761.1 | |
| NM_021131.4 | 5 | 481 | NP_066954.2 | |
| NM_178000.2 | 4 | 555 | NP_821067.1 | |
| NM_178001.2 | 5 | 660 | NP_821068.1 | |
| NM_178003.2 | NP_821070.1 | |||
| XM_011518834.2 | 5 | 480 | XP_011517136.1 | |
| XM_011518835.2 | 5 | 638 | XP_011517137.1 | |
| XM_011518836.2 | 4 | 394 | XP_011517138.1 | |
| XM_011518837.2 | 5 | 638 | XP_011517139.1 | |
| XM_011518838.2 | 4 | 375 | XP_011517140.1 | |
| XM_017014888.1 | 4 | 533 | XP_016870377.1 | |
| AK222788.1 | 4 | 454 | BAD96508.1 | |
| AK297202.1 | ||||
| AK302043.1 | 3 | 444 | ||
| AK302897.1 | 5 | 586 | ||
| AK307424.1 | 4 | 341 | ||
| AU125192.1 | ||||
| AU133279.1 | 4 | 551 | ||
| BC002545.2 | 5 | 465 | AAH02545.1 | |
| BC011605.2 | 5 | 465 | AAH11605.1 | |
| BE732116.1 | 5 | 540 | ||
| BG255640.1 | 4 | 440 | ||
| BT020119.1 | 4 | 268 | AAV38922.1 | |
| CD630703.1 | ||||
| CR749810.1 | CAH18670.1 | |||
| DA052599.1 | 4 | 341 | ||
| DA387148.1 | ||||
| DA737954.1 | 5 | 638 | ||
| DB009663.1 | 5 | 481 | ||
| X73478.1 | 4 | 457 | CAA51873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818725 | Chr.9:129026890 - 129176634 on Build GRCh38 | Gain |
|
| nsv831729 | Chr.9:128973042 - 129158330 on Build GRCh38 | Loss |
|
| nsv509323 | Chr.9:128991062 - 129129399 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map