Assay Details
Target Gene Details
Entrez Gene ID: | 222068 |
Gene Name: | transmembrane p24 trafficking protein 4 |
Gene Aliases: |
ERS25, GMP25iso, HNLF, p24a3, p24alpha3 |
Location: |
Chr.7:44577894-44582295 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMED4 | NM_001303058.1 | NP_001289987.1 | ||
| NM_001303059.1 | NP_001289988.1 | |||
| NM_001303060.1 | NP_001289989.1 | |||
| NM_001303061.1 | NP_001289990.1 | |||
| NM_001303062.1 | NP_001289991.1 | |||
| NM_182547.3 | NP_872353.2 | |||
| AK074557.1 | BAC11058.1 | |||
| AK290125.1 | ||||
| AK294122.1 | ||||
| AL602839.1 | ||||
| AY191223.1 | AAP20105.1 | |||
| BC052641.1 | AAH52641.1 | |||
| BC057851.1 | AAH57851.1 | |||
| BP872004.1 | ||||
| BX461603.2 | ||||
| BX647965.1 | ||||
| DA419871.1 |
More Information
Set Membership: |
|

Genomic Map