Assay Details
Target Gene Details
Entrez Gene ID: | 51744 |
Gene Name: | CD244 molecule |
Gene Aliases: |
2B4, NAIL, NKR2B4, Nmrk, SLAMF4 |
Location: |
Chr.1:160830160-160862902 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD244 | NM_001166663.1 | NP_001160135.1 | ||
| NM_001166664.1 | NP_001160136.1 | |||
| NM_016382.3 | NP_057466.1 | |||
| XM_011509621.2 | XP_011507923.1 | |||
| XM_011509622.2 | XP_011507924.1 | |||
| XM_011509623.2 | XP_011507925.1 | |||
| AF105261.1 | AAD32538.1 | |||
| AF107761.2 | AAD37838.1 | |||
| AF117711.1 | AAF28833.1 | |||
| AF145782.1 | AAD38951.1 | |||
| AF242540.1 | AAK00233.1 | |||
| AJ245375.1 | CAC00647.1 | |||
| AJ245376.1 | CAC00648.1 | |||
| AJ245377.1 | CAC00649.1 | |||
| AK313201.1 | ||||
| BC028073.1 | AAH28073.1 | |||
| BC053985.1 | AAH53985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548057 | Chr.1:160777825 - 160882256 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map