Assay Details
Target Gene Details
Entrez Gene ID: | 79178 |
Gene Name: | thiamine triphosphatase |
Gene Aliases: |
THTP, THTPASE |
Location: |
Chr.14:23511760-23559581 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THTPA | NR_046051.1 | |||
| NR_046052.1 | ||||
| AK090747.1 | ||||
| AK094211.1 | ||||
| BI488427.1 |
Target Gene Details
Entrez Gene ID: | 85446 |
Gene Name: | zinc finger homeobox 2 |
Gene Aliases: |
ZFH-5, ZNF409 |
Location: |
Chr.14:23520855-23555949 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFHX2 | NM_033400.2 | 9 | 4453 | NP_207646.2 |
| XM_005268135.3 | 5 | 1616 | XP_005268192.1 | |
| XM_011537245.2 | 9 | 4373 | XP_011535547.1 | |
| XM_011537246.2 | 9 | 6809 | XP_011535548.1 | |
| XM_011537247.2 | 9 | 5388 | XP_011535549.1 | |
| XM_011537252.2 | 5 | 1591 | XP_011535554.1 | |
| XM_017021714.1 | 9 | 4368 | XP_016877203.1 | |
| XM_017021715.1 | 9 | 4295 | XP_016877204.1 | |
| XM_017021716.1 | 8 | 4133 | XP_016877205.1 | |
| XM_017021718.1 | 6 | 1392 | XP_016877207.1 | |
| AB051549.1 | 1 | 761 | BAB21853.1 | |
| BC144336.1 | 1 | 958 | ||
| BC150606.1 | 1 | 958 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564043 | Chr.14:23495437 - 23550918 on Build GRCh38 | Loss |
|
| nsv952817 | Chr.14:23520192 - 23582591 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map