Assay Details
Target Gene Details
Entrez Gene ID: | 2847 |
Gene Name: | melanin concentrating hormone receptor 1 |
Gene Aliases: |
GPR24, MCH-1R, MCH1R, SLC-1, SLC1 |
Location: |
Chr.22:40679178-40682814 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCHR1 | NM_005297.3 | 2 | 2353 | NP_005288.3 |
| AF490537.2 | 2 | 2436 | AAO14670.1 | |
| AI934819.1 | 1 | 81 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955161 | Chr.22:40676797 - 40685096 on Build GRCh38 | Deletion |
|
| nsv834205 | Chr.22:40593419 - 40777442 on Build GRCh38 | Gain+Loss |
|
| nsv1160763 | Chr.22:40515733 - 40728482 on Build GRCh38 | Duplication |
|
| nsv1062438 | Chr.22:40646389 - 40806134 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map