Assay Details
Target Gene Details
Entrez Gene ID: | 867 |
Gene Name: | Cbl proto-oncogene |
Gene Aliases: |
C-CBL, CBL2, FRA11B, NSLL, RNF55 |
Location: |
Chr.11:119206276-119308149 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBL | NM_005188.3 | 2 | 422 | NP_005179.2 |
| XM_011543057.2 | 2 | 422 | XP_011541359.1 | |
| BC132733.1 | 2 | 351 | ||
| BC136463.1 | 2 | 351 | ||
| BG722940.1 | 2 | 412 | ||
| DB457271.2 | 2 | 422 | ||
| X57110.1 | 2 | 428 | CAA40393.1 |
More Information
Set Membership: |
|

Genomic Map