Assay Details
Target Gene Details
Entrez Gene ID: | 22841 |
Gene Name: | RAB11 family interacting protein 2 |
Gene Aliases: |
Rab11-FIP2, nRip11 |
Location: |
Chr.10:118004916-118046921 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB11FIP2 | NM_014904.2 | 5 | 1913 | NP_055719.1 |
| XM_005269629.3 | 6 | 2291 | XP_005269686.1 | |
| AB023158.1 | 5 | 1913 | ||
| AY037299.1 | 5 | 1913 | AAK68635.1 | |
| BC075073.2 | 5 | 1515 | AAH75073.1 | |
| BC075074.2 | 5 | 1515 | AAH75074.1 | |
| BX647717.1 | ||||
| DQ013303.1 | 6 | 1533 | AAY67796.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516870 | Chr.10:117945927 - 118028672 on Build GRCh38 | Loss |
|
| nsv1047815 | Chr.10:118004235 - 118069532 on Build GRCh38 | Gain |
|
| nsv832005 | Chr.10:117890359 - 118036955 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map