Assay Details
Target Gene Details
Entrez Gene ID: | 4682 |
Gene Name: | nucleotide binding protein 1 |
Gene Aliases: |
NBP, NBP1, NBP35 |
Location: |
Chr.16:10743164-10769351 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUBP1 | NM_001278506.1 | 10 | 1071 | NP_001265435.1 |
| NM_001323594.1 | 9 | 881 | NP_001310523.1 | |
| NM_001323595.1 | 10 | 946 | NP_001310524.1 | |
| NM_001323596.1 | 10 | 952 | NP_001310525.1 | |
| NM_001323597.1 | 10 | 1020 | NP_001310526.1 | |
| NM_002484.3 | 11 | 1104 | NP_002475.2 | |
| XM_017023252.1 | 10 | 1657 | XP_016878741.1 | |
| AK223204.1 | 11 | 1046 | BAD96924.1 | |
| BC100290.1 | 10 | 1014 | AAI00291.1 | |
| BC109322.1 | 11 | 1030 | AAI09323.1 | |
| BC109323.1 | 11 | 1030 | AAI09324.1 | |
| U01833.1 | 12 | 1030 | AAA61932.1 |
Target Gene Details
Entrez Gene ID: | 780776 |
Gene Name: | trans-golgi network vesicle protein 23 homolog A |
Gene Aliases: |
FAM18A, YDR084C |
Location: |
Chr.16:10758034-10818902 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TVP23A | NM_001079512.3 | NP_001072980.1 | ||
| NM_001318873.1 | NP_001305802.1 | |||
| NR_134901.1 | ||||
| NR_134902.1 | ||||
| NR_134903.1 | ||||
| NR_134904.1 | ||||
| NR_134905.1 | ||||
| NR_134906.1 | ||||
| XM_006720944.3 | XP_006721007.1 | |||
| XM_011522654.2 | 8 | 1800 | XP_011520956.1 | |
| XM_017023649.1 | 8 | 1362 | XP_016879138.1 | |
| XM_017023650.1 | 8 | 1562 | XP_016879139.1 | |
| XM_017023651.1 | 8 | 990 | XP_016879140.1 | |
| AK124106.1 | ||||
| AK294418.1 | ||||
| AL833564.1 | 6 | 765 | ||
| BC146882.1 | ||||
| BC146886.1 | ||||
| BC171817.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833140 | Chr.16:10664013 - 10791497 on Build GRCh38 | Loss |
|
| nsv1040428 | Chr.16:10145408 - 11362136 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map