Assay Details
Target Gene Details
Entrez Gene ID: | 392255 |
Gene Name: | growth differentiation factor 6 |
Gene Aliases: |
BMP-13, BMP13, CDMP2, KFM, KFS, KFS1, KFSL, LCA17, MCOP4, MCOPCB6, SCDO4, SGM1 |
Location: |
Chr.8:96142330-96160792 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDF6 | NM_001001557.2 | NP_001001557.1 | ||
| AJ537424.1 | CAD60934.1 |
More Information
Set Membership: |
|

Genomic Map