Assay Details
Target Gene Details
Entrez Gene ID: | 9456 |
Gene Name: | homer scaffolding protein 1 |
Gene Aliases: |
HOMER, HOMER1A, HOMER1B, HOMER1C, SYN47, Ves-1 |
Location: |
Chr.5:79373824-79514568 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOMER1 | NM_001277077.1 | NP_001264006.1 | ||
| NM_001277078.1 | NP_001264007.1 | |||
| NM_004272.4 | NP_004263.1 | |||
| AF093262.1 | AAC71026.1 | |||
| AK223471.1 | BAD97191.1 | |||
| AK312481.1 | ||||
| AY189939.1 | AAO38999.1 | |||
| AY189940.1 | AAO39000.1 | |||
| AY189941.1 | AAO39001.1 | |||
| BC015502.1 | AAH15502.1 | |||
| BC047438.1 | AAH47438.1 | |||
| BT009846.1 | AAP88848.1 | |||
| Y17829.1 | CAA76877.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890659 | Chr.5:79366031 - 79425620 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map