Assay Details
Target Gene Details
Entrez Gene ID: | 646626 |
Gene Name: | uncharacterized LOC646626 |
Gene Aliases: |
- |
Location: |
Chr.1:85276358-85278088 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC646626 | NR_045484.1 | 2 | 527 | |
| AK125723.1 | ||||
| BX104049.1 | 2 | 134 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34165 | Chr.1:85076357 - 85380566 on Build GRCh38 | Loss |
|
| nsv830459 | Chr.1:85123902 - 85320628 on Build GRCh38 | Loss |
|

Genomic Map