Assay Details
Target Gene Details
Entrez Gene ID: | 79707 |
Gene Name: | nucleolar protein 9 |
Gene Aliases: |
Grc3, NET6 |
Location: |
Chr.1:6521347-6554598 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOL9 | NM_024654.4 | 3 | 792 | NP_078930.3 |
| XM_005263493.3 | 2 | 180 | XP_005263550.1 | |
| XM_017002337.1 | 3 | 730 | XP_016857826.1 | |
| XM_017002338.1 | 3 | 535 | XP_016857827.1 | |
| XM_017002339.1 | 3 | 349 | XP_016857828.1 | |
| AK091284.1 | 3 | 729 | ||
| AK289653.1 | 3 | 724 | ||
| BC094836.1 | 2 | 167 | AAH94836.1 | |
| BC105095.1 | 3 | 792 | AAI05096.1 | |
| BC110849.1 | 3 | 701 | AAI10850.1 | |
| BC112278.1 | 3 | 792 | AAI12279.1 | |
| BI828678.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831647 | Chr.1:6525809 - 6587530 on Build GRCh38 | Loss |
|
| nsv7880 | Chr.1:6417968 - 6611914 on Build GRCh38 | Gain+Loss |
|
| nsv545268 | Chr.1:6490316 - 6578004 on Build GRCh38 | Loss |
|
| nsv545269 | Chr.1:6546548 - 6630080 on Build GRCh38 | Loss |
|
| nsv831536 | Chr.1:6429326 - 6605343 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map