Assay Details
Target Gene Details
Entrez Gene ID: | 56965 |
Gene Name: | poly(ADP-ribose) polymerase family member 6 |
Gene Aliases: |
ARTD17, PARP-6-B1, PARP-6-C, pART17 |
Location: |
Chr.15:72241181-72272575 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
More Information
Set Membership: |
|

Genomic Map