Assay Details
Target Gene Details
Entrez Gene ID: | 50512 |
Gene Name: | podocalyxin like 2 |
Gene Aliases: |
EG, PODLX2 |
Location: |
Chr.3:127629159-127672810 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PODXL2 | NM_015720.3 | 3 | 1083 | NP_056535.1 |
| AF219137.1 | 3 | 1042 | AAF44629.1 | |
| AY359096.1 | 3 | 1046 | AAQ89454.1 | |
| BC019330.1 | AAH19330.1 | |||
| BC052585.1 | 3 | 1038 | AAH52585.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8563n54 | Chr.3:127543914 - 127685471 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map