Assay Details
Target Gene Details
Entrez Gene ID: | 10561 |
Gene Name: | interferon induced protein 44 |
Gene Aliases: |
MTAP44, TLDC5, p44 |
Location: |
Chr.1:78648554-78664078 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFI44 | NM_006417.4 | NP_006408.3 | ||
| NR_135640.1 | ||||
| NR_135641.1 | ||||
| XM_005270380.3 | XP_005270437.1 | |||
| XM_011540516.2 | XP_011538818.1 | |||
| AI061108.1 | ||||
| BC022870.1 | AAH22870.1 | |||
| CA426666.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001291 | Chr.1:78598959 - 79684864 on Build GRCh38 | Loss |
|
| nsv830315 | Chr.1:78542758 - 78734906 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map