Assay Details
Target Gene Details
Entrez Gene ID: | 57561 |
Gene Name: | arrestin domain containing 3 |
Gene Aliases: |
TLIMP |
Location: |
Chr.5:91368724-91383359 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARRDC3 | NM_020801.2 | 1 | 447 | NP_065852.1 |
| AB037797.1 | 1 | 419 | ||
| AK291753.1 | 1 | 422 | ||
| BC015928.2 | 1 | 414 | AAH15928.1 | |
| BC053619.1 | 1 | 419 | AAH53619.1 | |
| DA110572.1 | 1 | 447 |
Target Gene Details
Entrez Gene ID: | 100129716 |
Gene Name: | ARRDC3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:91380347-91420715 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARRDC3-AS1 | NR_027435.1 |
More Information
Set Membership: |
|

Genomic Map