Assay Details
Target Gene Details
Entrez Gene ID: | 5426 |
Gene Name: | DNA polymerase epsilon, catalytic subunit |
Gene Aliases: |
CRCS12, FILS, POLE1 |
Location: |
Chr.12:132623762-132687524 on Build GRCh38 |
Assay Gene Location: | Within Exon 52 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLE | NM_006231.3 | 49 | 7811 | NP_006222.2 |
| AF128544.1 | 1 | 744 | ||
| AK025087.1 | 8 | 1884 | BAB15063.1 | |
| AK093003.1 | 12 | 2804 | ||
| AK128248.1 | 2 | 1902 | ||
| AL080203.1 | 5 | 1405 | CAB45774.1 | |
| BC007599.1 | 15 | 3137 | AAH07599.1 | |
| BC021559.1 | 10 | 2116 | AAH21559.1 | |
| BC087613.1 | 15 | 3137 | AAH87613.1 | |
| BX647647.1 | 23 | 5256 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952512 | Chr.12:132192256 - 132711214 on Build GRCh38 | Deletion |
|
| nsv561109 | Chr.12:132618549 - 132626173 on Build GRCh38 | Loss |
|
| esv2422284 | Chr.12:132604248 - 132708921 on Build GRCh38 | Deletion |
|
| nsv1037074 | Chr.12:132613063 - 132801953 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map