Assay Details
Target Gene Details
Entrez Gene ID: | 579 |
Gene Name: | NK3 homeobox 2 |
Gene Aliases: |
BAPX1, NKX3.2, NKX3B, SMMD |
Location: |
Chr.4:13540830-13544490 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX3-2 | NM_001189.3 | 2 | 870 | NP_001180.1 |
| AF009801.1 | 2 | 794 | AAB82783.1 | |
| BC111966.1 | 2 | 870 | AAI11967.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv593750 | Chr.4:13259081 - 13726709 on Build GRCh38 | Gain |
|
| nsv1014473 | Chr.4:11586127 - 13990352 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map