Assay Details
Target Gene Details
Entrez Gene ID: | 1314 |
Gene Name: | coatomer protein complex subunit alpha |
Gene Aliases: |
AILJK, HEP-COP |
Location: |
Chr.1:160288587-160343564 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPA | NM_001098398.1 | 1 | 381 | NP_001091868.1 |
| NM_004371.3 | 1 | 381 | NP_004362.2 | |
| AK307953.1 | 1 | 90 | ||
| BC038447.1 | 1 | 76 | AAH38447.1 | |
| DC308331.1 | 1 | 381 | ||
| U24105.1 | 2 | 453 | AAB70879.1 |
Target Gene Details
Entrez Gene ID: | 23385 |
Gene Name: | nicastrin |
Gene Aliases: |
ATAG1874 |
Location: |
Chr.1:160343056-160358952 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCSTN | XM_005245053.4 | 1 | 129 | XP_005245110.1 |
| XM_011509363.2 | 1 | 129 | XP_011507665.1 |
More Information
Set Membership: |
|

Genomic Map