Assay Details
Target Gene Details
Entrez Gene ID: | 5916 |
Gene Name: | retinoic acid receptor gamma |
Gene Aliases: |
NR1B3, RARC |
Location: |
Chr.12:53210566-53232256 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RARG | NM_000966.5 | NP_000957.1 | ||
| NM_001042728.2 | NP_001036193.1 | |||
| NM_001243730.1 | NP_001230659.1 | |||
| NM_001243731.1 | NP_001230660.1 | |||
| NM_001243732.1 | NP_001230661.1 | |||
| AB307689.1 | ||||
| AK290588.1 | ||||
| AK297023.1 | ||||
| AK297110.1 | ||||
| AK297277.1 | ||||
| AK297801.1 | ||||
| AK316259.1 | ||||
| BC093727.1 | AAH93727.1 | |||
| BC093729.1 | AAH93729.1 | |||
| HQ692828.1 | ||||
| HQ692829.1 | ||||
| L12060.1 | ||||
| M24857.1 | AAA52692.1 | |||
| M38258.1 | AAA60254.1 | |||
| M57707.1 | AAA63254.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832417 | Chr.12:53075874 - 53254231 on Build GRCh38 | Gain |
|
| nsv558994 | Chr.12:53103618 - 53277765 on Build GRCh38 | Loss |
|
| dgv2645n54 | Chr.12:53099603 - 53227927 on Build GRCh38 | Loss |
|
| nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss |
|
| nsv558996 | Chr.12:53211761 - 53289202 on Build GRCh38 | Loss |
|
| nsv469404 | Chr.12:53124036 - 53227927 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map