Assay Details
Target Gene Details
Entrez Gene ID: | 5866 |
Gene Name: | RAB3A interacting protein like 1 |
Gene Aliases: |
GRAB |
Location: |
Chr.11:61897296-61946312 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB3IL1 | NM_001271686.1 | 9 | 1299 | NP_001258615.1 |
| NM_013401.3 | 10 | 1542 | NP_037533.2 | |
| XM_005274140.4 | 10 | 1507 | XP_005274197.1 | |
| XM_006718647.3 | 10 | 1506 | XP_006718710.1 | |
| AF084557.1 | 9 | 1096 | AAF28399.1 | |
| BC022239.1 | 10 | 1458 | AAH22239.1 | |
| BC051820.1 | 9 | 1299 | AAH51820.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528697 | Chr.11:61690385 - 61919561 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map