Assay Details
Target Gene Details
Entrez Gene ID: | 55791 |
Gene Name: | ligand dependent nuclear receptor interacting factor 1 |
Gene Aliases: |
C1orf103, RIF1 |
Location: |
Chr.1:110877252-110964203 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRIF1 | NM_001006945.1 | 3 | 981 | NP_001006946.1 |
| NM_018372.3 | 4 | 2509 | NP_060842.3 | |
| XM_005271029.3 | 4 | 2765 | XP_005271086.1 | |
| XM_011541768.1 | 4 | 2509 | XP_011540070.1 | |
| XM_017001768.1 | 4 | 2506 | XP_016857257.1 | |
| XM_017001769.1 | XP_016857258.1 | |||
| AK001826.1 | 3 | 1087 | BAA91928.1 | |
| AK002131.1 | 3 | 1450 | BAA92097.1 | |
| AL834466.1 | 3 | 1427 | CAD39125.1 | |
| AY190122.1 | 4 | 2499 | AAO43631.1 | |
| BC008115.1 | 3 | 1651 | AAH08115.1 |
More Information
Set Membership: |
|

Genomic Map