Assay Details
Target Gene Details
Entrez Gene ID: | 391107 |
Gene Name: | olfactory receptor family 10 subfamily K member 2 |
Gene Aliases: |
OR1-4 |
Location: |
Chr.1:158419928-158420866 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10K2 | NM_001004476.1 | NP_001004476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831692 | Chr.1:158253669 - 158422226 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map