Assay Details
Target Gene Details
Entrez Gene ID: | 84909 |
Gene Name: | chromosome 9 open reading frame 3 |
Gene Aliases: |
AOPEP, AP-O, APO, C90RF3, ONPEP |
Location: |
Chr.9:94726604-95148264 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C9orf3 | XM_011519121.2 | XP_011517423.1 |
Target Gene Details
Entrez Gene ID: | 2176 |
Gene Name: | Fanconi anemia complementation group C |
Gene Aliases: |
FA3, FAC, FACC |
Location: |
Chr.9:95099054-95317730 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FANCC | NM_000136.2 | NP_000127.2 | ||
| NM_001243743.1 | NP_001230672.1 | |||
| XM_005251802.3 | XP_005251859.1 | |||
| XM_006717001.2 | XP_006717064.1 | |||
| XM_011518365.2 | XP_011516667.1 | |||
| XM_011518367.2 | XP_011516669.1 | |||
| XM_017014452.1 | XP_016869941.1 | |||
| XM_017014453.1 | XP_016869942.1 | |||
| XM_017014454.1 | XP_016869943.1 | |||
| AK312548.1 | ||||
| BC015748.1 | AAH15748.1 | |||
| CB052780.1 | ||||
| KU178051.1 | ||||
| KU178052.1 | ||||
| X66893.1 | CAA47347.1 | |||
| X66894.1 | CAA47348.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038106 | Chr.9:95030756 - 95119134 on Build GRCh38 | Gain |
|
| nsv831656 | Chr.9:95069269 - 95227698 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map