Assay Details
Target Gene Details
Entrez Gene ID: | 3229 |
Gene Name: | homeobox C13 |
Gene Aliases: |
ECTD9, HOX3, HOX3G |
Location: |
Chr.12:53938792-53946544 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXC13 | NM_017410.2 | 2 | 2020 | NP_059106.2 |
| AA722686.1 | 1 | 379 | ||
| AF255676.1 | 2 | 2020 | AAF67760.1 | |
| AF263466.2 | 2 | 1977 | AAF73439.1 | |
| AK024027.1 | 2 | 1502 | BAB14786.1 | |
| BC090850.1 | 2 | 1989 | AAH90850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826379 | Chr.12:53935085 - 54056069 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114528288] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map