Assay Details
Target Gene Details
Entrez Gene ID: | 54974 |
Gene Name: | tRNA-histidine guanylyltransferase 1 like |
Gene Aliases: |
ICF45, IHG-1, hTHG1 |
Location: |
Chr.5:157731389-157741449 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THG1L | NM_001317824.1 | NP_001304753.1 | ||
| NM_001317825.1 | NP_001304754.1 | |||
| NM_001317826.1 | NP_001304755.1 | |||
| NM_017872.4 | NP_060342.2 | |||
| XM_017009603.1 | XP_016865092.1 | |||
| AF086486.1 | ||||
| AK000553.1 | BAA91249.1 | |||
| AK021663.1 | BAB13870.1 | |||
| AK023355.1 | BAB14540.1 | |||
| AK223119.1 | BAD96839.1 | |||
| AK304590.1 | ||||
| AL136669.1 | CAB66604.1 | |||
| AY463216.1 | AAS21134.1 | |||
| BC001523.2 | AAH01523.2 | |||
| BC001852.2 | AAH01852.2 | |||
| BC023521.2 | AAH23521.1 | |||
| CR533503.1 | CAG38534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759389 | Chr.5:157570600 - 157806036 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map