Assay Details
Target Gene Details
Entrez Gene ID: | 3612 |
Gene Name: | inositol monophosphatase 1 |
Gene Aliases: |
IMP, IMPA |
Location: |
Chr.8:81656916-81686354 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IMPA1 | NM_001144878.1 | 9 | 995 | NP_001138350.1 |
| NM_001144879.1 | 7 | 685 | NP_001138351.1 | |
| NM_005536.3 | 8 | 794 | NP_005527.1 | |
| AF042729.2 | 8 | 765 | AAB97468.1 | |
| AK297078.1 | 7 | 632 | ||
| AK300750.1 | 9 | 924 | ||
| AK312823.1 | 8 | 740 | ||
| BC008381.1 | 8 | 760 | AAH08381.1 | |
| BC009565.1 | 8 | 764 | AAH09565.1 | |
| X66922.1 | 7 | 703 | CAA47359.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611607 | Chr.8:81405621 - 81768032 on Build GRCh38 | Gain |
|
| nsv831369 | Chr.8:81605833 - 81823586 on Build GRCh38 | Gain+Loss |
|
| nsv831368 | Chr.8:81554926 - 81697395 on Build GRCh38 | Gain |
|
| esv2737230 | Chr.8:81633446 - 81689714 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map