Assay Details
Target Gene Details
Entrez Gene ID: | 159686 |
Gene Name: | cilia and flagella associated protein 58 |
Gene Aliases: |
C10orf80, CCDC147, bA127L20.4, bA127L20.5, bA554P13.1 |
Location: |
Chr.10:104339124-104455090 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP58 | NM_001008723.1 | 5 | 823 | NP_001008723.1 |
| XM_011539377.2 | 5 | 1542 | XP_011537679.1 | |
| XM_017015787.1 | 5 | 1194 | XP_016871276.1 | |
| XM_017015788.1 | 5 | 1725 | XP_016871277.1 | |
| XM_017015789.1 | 5 | 812 | XP_016871278.1 | |
| XM_017015790.1 | 5 | 812 | XP_016871279.1 | |
| AK093227.1 | 5 | 811 | BAC04102.1 | |
| AK296463.1 | 5 | 816 | ||
| BC036225.1 | 5 | 825 | AAH36225.1 | |
| BC109126.2 | 5 | 777 | AAI09127.1 | |
| BX641120.1 | 5 | 822 |
More Information
Set Membership: |
|

Genomic Map