Assay Details
Target Gene Details
Entrez Gene ID: | 55735 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C11 |
Gene Aliases: |
dJ126A5.1 |
Location: |
Chr.1:6634168-6701906 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC11 | NM_018198.3 | 8 | 958 | NP_060668.2 |
| XM_017001745.1 | 7 | 811 | XP_016857234.1 | |
| AF306694.1 | 8 | 857 | ||
| AF306695.1 | 8 | 872 | ||
| AK001599.1 | 8 | 867 | BAA91780.1 | |
| AK294539.1 | 7 | 781 | ||
| AK298378.1 | 8 | 844 | ||
| AL109978.1 | CAB53376.1 | |||
| AL833841.1 | 8 | 832 | CAD38701.1 | |
| AY336749.1 | 8 | 868 | AAR02411.1 | |
| BC006086.1 | 8 | 854 | AAH06086.1 | |
| BC008772.1 | 8 | 859 | AAH08772.1 | |
| BC014145.2 | 8 | 866 | AAH14145.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv437199 | Chr.1:6642091 - 6743687 on Build GRCh38 | Loss |
|
| nsv1004598 | Chr.1:6645296 - 7178603 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map