Assay Details
Target Gene Details
Entrez Gene ID: | 89894 |
Gene Name: | transmembrane protein 116 |
Gene Aliases: |
- |
Location: |
Chr.12:111894936-112013219 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044606 | Chr.12:111885161 - 111989037 on Build GRCh38 | Gain |
|
| nsv1045310 | Chr.12:111927424 - 112012015 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map