Assay Details
Target Gene Details
Entrez Gene ID: | 54101 |
Gene Name: | receptor interacting serine/threonine kinase 4 |
Gene Aliases: |
ANKK2, ANKRD3, DIK, NKRD3, PKK, PPS2, RIP4 |
Location: |
Chr.21:41739369-41767089 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIPK4 | NM_020639.2 | 8 | 2086 | NP_065690.2 |
| AB047783.1 | 8 | 2062 | BAB56136.1 | |
| AF308292.1 | 2 | 1101 | AAG48260.1 | |
| AJ278016.1 | 8 | 2086 | CAC04247.1 | |
| AK027424.1 | 8 | 2103 | BAB55102.1 | |
| AK296685.1 | 6 | 1203 | ||
| AK302592.1 | 8 | 1897 | ||
| AK316412.1 | 8 | 2049 | ||
| AL137448.1 | 1 | 879 | CAB70741.2 | |
| BC110617.1 | 8 | 2059 | AAI10618.1 | |
| BC110618.1 | 8 | 2059 | AAI10619.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893416 | Chr.21:41494369 - 41860297 on Build GRCh38 | Gain |
|
| nsv1065094 | Chr.21:41712854 - 41794037 on Build GRCh38 | Loss |
|
| nsv459265 | Chr.21:41732924 - 41760723 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map