Assay Details
Target Gene Details
Entrez Gene ID: | 6139 |
Gene Name: | ribosomal protein L17 |
Gene Aliases: |
L17, PD-1, RPL23 |
Location: |
Chr.18:49488481-49492565 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL17 | NM_000985.4 | NP_000976.1 | ||
| NM_001035006.2 | NP_001030178.1 | |||
| NM_001199340.1 | NP_001186269.1 | |||
| NM_001199341.1 | NP_001186270.1 | |||
| NM_001199342.1 | NP_001186271.1 | |||
| NM_001199343.1 | NP_001186272.1 | |||
| NM_001199344.1 | NP_001186273.1 | |||
| NM_001199345.1 | NP_001186274.1 | |||
| AK304659.1 | ||||
| AK311828.1 | ||||
| BG497914.1 | ||||
| BI086293.1 | ||||
| CD386294.1 | ||||
| CD387752.1 |
Target Gene Details
Entrez Gene ID: | 100526842 |
Gene Name: | RPL17-C18orf32 readthrough |
Gene Aliases: |
- |
Location: |
Chr.18:49481178-49492565 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL17-C18orf32 | NM_001199356.1 | NP_001186285.1 | ||
| BI461805.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2294 | Chr.18:49470841 - 49492546 on Build GRCh38 | Deletion |
|
| nsv1065279 | Chr.18:49468523 - 49512116 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map