Assay Details
Target Gene Details
Entrez Gene ID: | 6521 |
Gene Name: | solute carrier family 4 member 1 (Diego blood group) |
Gene Aliases: |
AE1, BND3, CD233, CHC, DI, EMPB3, EPB3, FR, RTA1A, SAO, SPH4, SW, WD, WD1, WR |
Location: |
Chr.17:44248390-44268161 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC4A1 | NM_000342.3 | 20 | 2950 | NP_000333.1 |
| XM_005257593.4 | 18 | 2718 | XP_005257650.1 | |
| XM_011525129.1 | 19 | 2887 | XP_011523431.1 | |
| BC096106.3 | 19 | 2863 | AAH96106.1 | |
| BC096107.3 | 19 | 2863 | AAH96107.1 | |
| BC099628.3 | 19 | 2863 | AAH99628.1 | |
| BC099629.3 | 19 | 2863 | AAH99629.1 | |
| BC101570.1 | AAI01571.1 | |||
| BC101574.1 | AAI01575.1 | |||
| DQ419529.1 | ABD74692.1 | |||
| GQ981383.1 | 19 | 2844 | ||
| GQ981384.1 | 19 | 2844 | ||
| GQ981385.1 | 14 | 1746 | ||
| GQ981386.1 | 19 | 2844 | ||
| GQ981387.1 | 19 | 2844 | ||
| M27819.1 | 20 | 2915 | AAA35514.1 | |
| X12609.1 | 20 | 2951 | CAA31128.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510713 | Chr.17:44201179 - 44340387 on Build GRCh38 | Deletion |
|
| nsv833455 | Chr.17:44108553 - 44297300 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map