Assay Details
Target Gene Details
Entrez Gene ID: | 59336 |
Gene Name: | PR domain 13 |
Gene Aliases: |
MU-MB-20.220, PFM10 |
Location: |
Chr.6:99606774-99615578 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRDM13 | NM_021620.3 | 4 | 1999 | NP_067633.2 |
| AY004253.1 | 4 | 2029 | AAG13448.1 | |
| AY130010.1 | 1 | 242 | AAN05093.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830741 | Chr.6:99611699 - 99808129 on Build GRCh38 | Gain |
|
| nsv1029536 | Chr.6:98422726 - 99818247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map