Assay Details
Target Gene Details
Entrez Gene ID: | 59271 |
Gene Name: | eva-1 homolog C (C. elegans) |
Gene Aliases: |
B18, B19, C21orf63, C21orf64, FAM176C, PRED34, SUE21 |
Location: |
Chr.21:32411692-32515400 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EVA1C | NM_001286556.1 | NP_001273485.1 | ||
| NM_001320744.1 | NP_001307673.1 | |||
| NM_001320745.1 | NP_001307674.1 | |||
| NM_058187.4 | NP_478067.2 | |||
| NR_104472.1 | ||||
| NR_135467.1 | ||||
| XM_005261019.4 | XP_005261076.1 | |||
| XM_006724038.3 | XP_006724101.1 | |||
| XM_017028418.1 | XP_016883907.1 | |||
| XM_017028419.1 | XP_016883908.1 | |||
| XM_017028420.1 | XP_016883909.1 | |||
| XM_017028421.1 | XP_016883910.1 | |||
| XM_017028422.1 | XP_016883911.1 | |||
| XM_017028423.1 | XP_016883912.1 | |||
| AF358258.1 | AAL40411.1 | |||
| AL543462.3 | ||||
| AY040087.1 | AAK74135.1 | |||
| AY358787.1 | AAQ89147.1 | |||
| BC038710.1 | AAH38710.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834085 | Chr.21:32322748 - 32519107 on Build GRCh38 | Gain |
|
| nsv3494 | Chr.21:32401757 - 32436315 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map