Assay Details
Target Gene Details
Entrez Gene ID: | 2275 |
Gene Name: | four and a half LIM domains 3 |
Gene Aliases: |
SLIM2 |
Location: |
Chr.1:37996765-38005515 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FHL3 | NM_001243878.1 | 5 | 1221 | NP_001230807.1 |
| NM_004468.4 | 6 | 1397 | NP_004459.2 | |
| XM_017000675.1 | 3 | 821 | XP_016856164.1 | |
| AF133732.1 | 5 | 1218 | AAF61376.1 | |
| BC001351.2 | 6 | 1387 | AAH01351.1 | |
| BC011697.2 | 6 | 1387 | AAH11697.1 | |
| BC014501.1 | 5 | 1208 | AAH14501.1 | |
| U60116.2 | 5 | 1257 | AAC04466.2 |
More Information
Set Membership: |
|

Genomic Map