Assay Details
Target Gene Details
Entrez Gene ID: | 9677 |
Gene Name: | diphosphoinositol pentakisphosphate kinase 1 |
Gene Aliases: |
HISPPD2A, IP6K, IPS1, VIP1, hsVIP1 |
Location: |
Chr.15:43533462-43590272 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 31 - Exon 32 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832988 | Chr.15:43512443 - 43684664 on Build GRCh38 | Loss |
|
| nsv437799 | Chr.15:43491802 - 43697320 on Build GRCh38 | Loss |
|
| nsv428304 | Chr.15:43530960 - 43716382 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map