Assay Details
Target Gene Details
Entrez Gene ID: | 80122 |
Gene Name: | mitogen-activated protein kinase kinase kinase 19 |
Gene Aliases: |
RCK, YSK4 |
Location: |
Chr.2:134963875-135047468 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K19 | NM_001018044.2 | 7 | 3274 | NP_001018054.1 |
| NM_001018046.2 | 8 | 1159 | NP_001018056.1 | |
| NM_001018047.2 | 8 | 1015 | NP_001018057.1 | |
| NM_001282883.1 | 7 | 1009 | NP_001269812.1 | |
| NM_001321177.1 | 10 | 3905 | NP_001308106.1 | |
| NM_025052.4 | 9 | 3613 | NP_079328.3 | |
| XM_011511891.2 | 9 | 3753 | XP_011510193.1 | |
| XM_011511892.1 | 12 | 4321 | XP_011510194.1 | |
| XM_011511894.2 | 9 | 3728 | XP_011510196.1 | |
| XM_011511896.2 | 9 | 3594 | XP_011510198.1 | |
| XM_011511897.2 | 9 | 3674 | XP_011510199.1 | |
| XM_017005003.1 | 12 | 4288 | XP_016860492.1 | |
| XM_017005004.1 | 11 | 4173 | XP_016860493.1 | |
| XM_017005005.1 | 10 | 3974 | XP_016860494.1 | |
| XM_017005006.1 | 10 | 3336 | XP_016860495.1 | |
| XM_017005007.1 | 9 | 3721 | XP_016860496.1 | |
| AK026727.1 | 1 | 494 | BAB15538.1 | |
| AK122935.1 | 10 | 3905 | ||
| AK131216.1 | 3 | 1752 | ||
| AK225579.1 | 1 | 494 | ||
| AY574901.1 | 9 | 3613 | AAT81410.1 | |
| AY574902.1 | 7 | 3274 | AAT81411.1 | |
| AY574903.1 | AAT81412.1 | |||
| AY574904.1 | 8 | 1159 | AAT81413.1 | |
| AY574905.1 | 8 | 1015 | AAT81414.1 | |
| BC034417.1 | 2 | 660 | AAH34417.1 | |
| BC137276.1 | 9 | 3612 | ||
| BC137277.1 | 9 | 3612 | ||
| BC144555.1 | 7 | 1008 |
More Information
Set Membership: |
|

Genomic Map