Assay Details
Target Gene Details
Entrez Gene ID: | 92002 |
Gene Name: | family with sequence similarity 58 member A |
Gene Aliases: |
STAR |
Location: |
Chr.X:153587925-153599177 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM58A | NM_001130997.2 | NP_001124469.1 | ||
| NM_152274.4 | NP_689487.2 | |||
| XM_005277920.4 | XP_005277977.1 | |||
| XM_005277921.4 | XP_005277978.1 | |||
| XM_011531214.2 | XP_011529516.1 | |||
| XM_011531215.2 | XP_011529517.1 | |||
| AL549300.3 | ||||
| BC001909.1 | ||||
| BC007232.1 | ||||
| BC032121.2 | AAH32121.1 | |||
| BC071851.1 | AAH71851.1 | |||
| DQ323993.1 | ABC88595.1 | |||
| DT217835.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map