Assay Details
Target Gene Details
Entrez Gene ID: | 54541 |
Gene Name: | DNA damage inducible transcript 4 |
Gene Aliases: |
Dig2, REDD-1, REDD1 |
Location: |
Chr.10:72273919-72276039 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDIT4 | NM_019058.2 | NP_061931.1 | ||
| AF335324.1 | AAL38424.1 | |||
| AK000507.1 | BAA91214.1 | |||
| AL136668.1 | CAB66603.1 | |||
| BC000708.1 | AAH00708.1 | |||
| BC007714.1 | AAH07714.1 | |||
| BC015236.1 | AAH15236.1 | |||
| BM843387.1 | ||||
| CN428398.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761604 | Chr.10:72262756 - 72287410 on Build GRCh38 | Loss |
|
| nsv551514 | Chr.10:72243964 - 72286159 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map