Assay Details
Target Gene Details
Entrez Gene ID: | 51702 |
Gene Name: | peptidyl arginine deiminase 3 |
Gene Aliases: |
PAD3, PDI3 |
Location: |
Chr.1:17249079-17284233 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PADI3 | NM_016233.2 | NP_057317.2 | ||
| XM_011541571.2 | XP_011539873.1 | |||
| XM_011541572.2 | XP_011539874.1 | |||
| AB026831.1 | BAA85974.1 | |||
| BC041592.1 | AAH41592.1 | |||
| BC109091.1 | AAI09092.1 | |||
| BC109092.1 | AAI09093.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv128n100 | Chr.1:17249833 - 17309505 on Build GRCh38 | Gain |
|
| nsv834291 | Chr.1:17204322 - 17350217 on Build GRCh38 | Gain |
|
| nsv521814 | Chr.1:17260158 - 17265829 on Build GRCh38 | Loss |
|
| dgv129n100 | Chr.1:17261667 - 17296402 on Build GRCh38 | Gain |
|
| nsv508958 | Chr.1:17244847 - 17296637 on Build GRCh38 | Insertion |
|
| nsv545692 | Chr.1:17228358 - 17331039 on Build GRCh38 | Loss |
|
| nsv470700 | Chr.1:17225448 - 17268097 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map