Assay Details
Target Gene Details
Entrez Gene ID: | 374403 |
Gene Name: | TBC1 domain family member 10C |
Gene Aliases: |
CARABIN, EPI64C |
Location: |
Chr.11:67403718-67410090 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D10C | NM_001256508.1 | NP_001243437.1 | ||
| NM_198517.3 | NP_940919.1 | |||
| NR_046266.1 | ||||
| XM_006718538.3 | XP_006718601.1 | |||
| XM_006718539.3 | XP_006718602.1 | |||
| XM_006718540.3 | XP_006718603.1 | |||
| XM_006718541.3 | XP_006718604.1 | |||
| XM_006718542.3 | XP_006718605.1 | |||
| XM_006718543.2 | XP_006718606.1 | |||
| XM_006718544.3 | XP_006718607.1 | |||
| XM_011545001.2 | XP_011543303.1 | |||
| XM_011545002.2 | XP_011543304.1 | |||
| AK122585.1 | 2 | 608 | BAC56926.1 | |
| AK126131.1 | 1 | 2014 | ||
| BC036873.1 | AAH36873.3 | |||
| BC062999.1 | AAH62999.1 | |||
| CR979868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss |
|
| dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map