Assay Details
Target Gene Details
Entrez Gene ID: | 84296 |
Gene Name: | GINS complex subunit 4 |
Gene Aliases: |
SLD5 |
Location: |
Chr.8:41529206-41545046 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GINS4 | NM_032336.2 | NP_115712.1 | ||
| XM_005273659.3 | XP_005273716.1 | |||
| AK095334.1 | ||||
| AK299207.1 | ||||
| AK313373.1 | ||||
| BC005995.1 | AAH05995.1 | |||
| BC027454.1 | AAH27454.1 | |||
| DC376966.1 |
Target Gene Details
Entrez Gene ID: | 102723729 |
Gene Name: | uncharacterized LOC102723729 |
Gene Aliases: |
- |
Location: |
Chr.8:41534305-41578368 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102723729 | NR_125824.1 | |||
| AI702202.1 | ||||
| BE326543.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831299 | Chr.8:41448617 - 41692486 on Build GRCh38 | Loss |
|
| esv3576565 | Chr.8:41064045 - 41790840 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map