Assay Details
Target Gene Details
Entrez Gene ID: | 25999 |
Gene Name: | CAP-Gly domain containing linker protein 3 |
Gene Aliases: |
CLIPR-59, CLIPR59, RSNL1 |
Location: |
Chr.19:36014660-36032895 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLIP3 | NM_001199570.1 | NP_001186499.1 | ||
| NM_015526.2 | NP_056341.1 | |||
| AJ427922.1 | CAD20873.1 | |||
| AK056445.1 | ||||
| AK090732.1 | ||||
| AK092354.1 | ||||
| AK094738.1 | ||||
| AK289509.1 | ||||
| AK294748.1 | ||||
| BC013116.1 | AAH13116.2 | |||
| DA398172.1 | ||||
| DA406432.1 |
Target Gene Details
Entrez Gene ID: | 101927572 |
Gene Name: | uncharacterized LOC101927572 |
Gene Aliases: |
- |
Location: |
Chr.19:36014499-36045997 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927572 | NM_001290056.1 | NP_001276985.1 |
More Information
Set Membership: |
|

Genomic Map