Assay Details
Target Gene Details
Entrez Gene ID: | 64857 |
Gene Name: | pleckstrin homology and RhoGEF domain containing G2 |
Gene Aliases: |
ARHGEF42, CLG, LDAMD |
Location: |
Chr.19:39412582-39428415 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHG2 | NM_022835.2 | 18 | 2080 | NP_073746.2 |
| XM_005259163.2 | 18 | 1901 | XP_005259220.1 | |
| XM_006723334.2 | 18 | 2431 | XP_006723397.1 | |
| XM_011527232.2 | 18 | 2007 | XP_011525534.1 | |
| XM_017027150.1 | 17 | 2572 | XP_016882639.1 | |
| XM_017027151.1 | 17 | 2563 | XP_016882640.1 | |
| AK024429.1 | 18 | 2080 | ||
| AK026111.1 | BAB15364.1 | |||
| AK302476.1 | 18 | 1704 | ||
| BC013426.1 | 8 | 571 | ||
| BC075858.1 | 14 | 1358 | AAH75858.1 |
More Information
Set Membership: |
|

Genomic Map