Assay Details
Target Gene Details
Entrez Gene ID: | 29895 |
Gene Name: | myosin light chain, phosphorylatable, fast skeletal muscle |
Gene Aliases: |
HUMMLC2B, MLC2B, MRLC2, MYL11 |
Location: |
Chr.16:30370934-30377991 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYLPF | NM_001324458.1 | NP_001311387.1 | ||
| NM_001324459.1 | NP_001311388.1 | |||
| NM_013292.4 | NP_037424.2 | |||
| AF363061.1 | AAK52797.1 | |||
| AK314082.1 | ||||
| BC012571.1 | AAH12571.1 | |||
| BI828249.1 | ||||
| BI837593.1 | ||||
| BX649054.1 | ||||
| CR456963.1 | CAG33244.1 | |||
| M21812.1 | AAA91848.1 | |||
| X57543.1 | CAA40762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827612 | Chr.16:30360217 - 30418541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map