Assay Details
Target Gene Details
Entrez Gene ID: | 1572 |
Gene Name: | cytochrome P450 family 2 subfamily F member 1 |
Gene Aliases: |
C2F1, CYP2F, CYPIIF1 |
Location: |
Chr.19:41114292-41128381 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP2F1 | NM_000774.4 | NP_000765.2 | ||
| NR_135528.1 | ||||
| XM_011526551.2 | XP_011524853.1 | |||
| XM_011526552.2 | XP_011524854.1 | |||
| XM_011526553.2 | XP_011524855.1 | |||
| XM_011526554.2 | XP_011524856.1 | |||
| XM_011526555.2 | XP_011524857.1 | |||
| XM_017026384.1 | XP_016881873.1 | |||
| XM_017026385.1 | XP_016881874.1 | |||
| XM_017026386.1 | XP_016881875.1 | |||
| XM_017026387.1 | XP_016881876.1 | |||
| AK296884.1 | ||||
| AK298082.1 | ||||
| AK316498.1 | ||||
| BC109056.1 | AAI09057.1 | |||
| DC413790.1 | ||||
| J02906.1 | AAA52156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2492 | Chr.19:41096554 - 41131195 on Build GRCh38 | Insertion |
|
| esv3644372 | Chr.19:41119456 - 41147784 on Build GRCh38 | Gain |
|
| nsv1065684 | Chr.19:41010484 - 41138637 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map