Assay Details
Target Gene Details
Entrez Gene ID: | 23409 |
Gene Name: | sirtuin 4 |
Gene Aliases: |
SIR2L4 |
Location: |
Chr.12:120292199-120313242 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRT4 | NM_012240.2 | 3 | 704 | NP_036372.1 |
| XM_005253865.4 | 3 | 432 | XP_005253922.1 | |
| XM_006719308.2 | 3 | 1390 | XP_006719371.1 | |
| XM_006719309.3 | 3 | 886 | XP_006719372.1 | |
| AF083109.1 | 3 | 665 | AAD40852.1 | |
| BC109319.3 | 3 | 665 | AAI09320.1 | |
| BC109320.3 | 3 | 665 | AAI09321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763006 | Chr.12:120293543 - 120312943 on Build GRCh38 | Loss |
|
| nsv832530 | Chr.12:120148265 - 120320372 on Build GRCh38 | Loss |
|
| nsv518764 | Chr.12:120310110 - 120312714 on Build GRCh38 | Loss |
|
| nsv560435 | Chr.12:120257338 - 120331854 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map